3-(Aminomethyl)benzenesulphonamide

Lieferant: Apollo Scientific
Gefahr

OR52244-1G OR52244-250MG
APOSOR52244-1GEA 590 EUR
APOSOR52244-1G APOSOR52244-250MG
3-(Aminomethyl)benzenesulphonamide
3-(Aminomethyl)benzenesulphonamide
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186.232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09727958
CAS-Nummer: 628298-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR