Oxan-3-amine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Tetrahydro-2H-pyran-3-amine
OR52254-1G
OR52254-250MG
APOSOR52254-1GEA
0
EUR
APOSOR52254-1G
APOSOR52254-250MG
Oxan-3-amine
Oxan-3-amine
Formel:
C₅H₁₁NO Molecular Weight: 101,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371531 CAS-Nummer: 120811-32-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...