Cyclopropanecarboxylic acid anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cyclopropanecarboxylic anhydride
OR52359-10G
OR52359-1G
OR52359-5G
APOSOR52359-10GEA
742
EUR
APOSOR52359-10G
APOSOR52359-1G
APOSOR52359-5G
Cyclopropanecarboxylic acid anhydride
Cyclopropanecarboxylic acid anhydride
Formel:
C₈H₁₀O₃ Molecular Weight: 154,163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12404941 CAS-Nummer: 33993-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...