2-Amino-6-bromobenzaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-2-formylaniline
OR52365-1G
OR52365-5G
APOSOR52365-1GEA
212
EUR
APOSOR52365-1G
APOSOR52365-5G
2-Amino-6-bromobenzaldehyde 98%
2-Amino-6-bromobenzaldehyde
Formel:
C₇H₆BrNO Molecular Weight: 200,033 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11036195 CAS-Nummer: 135484-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...