4-Amino-1-methylpyridin-2-one hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-1-methylpyridin-2(1H)-one hydrochloride
OR52375-1G
OR52375-5G
APOSOR52375-1GEA
168
EUR
APOSOR52375-1G
APOSOR52375-5G
4-Amino-1-methylpyridin-2-one hydrochloride
4-Amino-1-methylpyridin-2-one hydrochloride
Formel:
C₆H₈N₂O·ClH Molecular Weight: 160,601 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20278328 CAS-Nummer: 1404373-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...