4-(Bromomethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: [4-(Bromomethyl)phenyl]methanol, (4-(Bromomethyl)phenyl)methanol

OR52377-10G OR52377-1G OR52377-5G
APOSOR52377-10GEA 368 EUR
APOSOR52377-10G APOSOR52377-1G APOSOR52377-5G
4-(Bromomethyl)benzyl alcohol 97%
4-(Bromomethyl)benzyl alcohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
Schmelzpunkt: 76…77 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08275218
CAS-Nummer: 71831-21-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR