4-Iodo-5-methyl-1H-imidazole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Iodo-4-methyl-imidazole

OR52397-1G OR52397-5G OR52397-25G
APOSOR52397-1GEA 96 EUR
APOSOR52397-1G APOSOR52397-5G APOSOR52397-25G
4-Iodo-5-methyl-1H-imidazole 97%
4-Iodo-5-methyl-1H-imidazole
Formel: C₄H₅IN₂
Molecular Weight: 208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24396002
CAS-Nummer: 15813-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR