4,6-Dichloro-2,5-dimethylpyrimidine 99%

Lieferant: Apollo Scientific
Achtung

OR52409-10G OR52409-1G OR52409-5G
APOSOR52409-10GEA 120 EUR
APOSOR52409-10G APOSOR52409-1G APOSOR52409-5G
4,6-Dichloro-2,5-dimethylpyrimidine 99%
4,6-Dichloro-2,5-dimethylpyrimidine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,031 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09991674
CAS-Nummer: 1780-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR