N-Cyano-N-phenyl-p-toluenesulphonamide
Lieferant: Apollo Scientific
Achtung
Synonyme: N-Cyano-4-methyl-N-phenylbenzenesulphonamide
OR52467-1G OR52467-5G
APOSOR52467-1GEA 190 EUR
APOSOR52467-1G APOSOR52467-5G
N-Cyano-N-phenyl-p-toluenesulphonamide
N-Cyano-N-phenyl-p-toluenesulphonamide
Formel: C₁₄H₁₂N₂O₂S
Molecular Weight: 272.322 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00159358
CAS-Nummer: 55305-43-6