5-[4-(Propan-2-yl)piperazin-1-yl]pyridin-2-amine

Lieferant: Apollo Scientific

Synonyme: 5-(4-Isopropylpiperazin-1-yl)pyridin-2-amine

OR52472-100MG OR52472-1G OR52472-250MG
APOSOR52472-100MGEA 78.3 EUR
APOSOR52472-100MG APOSOR52472-1G APOSOR52472-250MG
5-[4-(Propan-2-yl)piperazin-1-yl]pyridin-2-amine
5-[4-(Propan-2-yl)piperazin-1-yl]pyridin-2-amine
Formel: C₁₂H₂₀N₄
Molecular Weight: 220,314 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12175168
CAS-Nummer: 866620-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR