2-Bromo-4-chloropyrimidine

Lieferant: Apollo Scientific
Achtung

OR52503-1G OR52503-100MG OR52503-250MG
APOSOR52503-1GEA 364 EUR
APOSOR52503-1G APOSOR52503-100MG APOSOR52503-250MG
2-Bromo-4-chloropyrimidine
2-Bromo-4-chloropyrimidine
Formel: C₄H₂BrClN₂
Molecular Weight: 193,429 g/mol
Schmelzpunkt: 47…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847634
CAS-Nummer: 885702-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR