2-Bromo-4,6-dichloropyrimidine

Lieferant: Apollo Scientific
Achtung

OR52504-1G OR52504-100MG OR52504-250MG
APOSOR52504-1GEA 406 EUR
APOSOR52504-1G APOSOR52504-100MG APOSOR52504-250MG
2-Bromo-4,6-dichloropyrimidine
2-Bromo-4,6-dichloropyrimidine
Formel: C₄HBrCl₂N₂
Molecular Weight: 227,874 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566618
CAS-Nummer: 1399480-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR