2-Carboxypyridine-4-boronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxylic acid

OR52669-1G OR52669-250MG
APOSOR52669-1GEA 821 EUR
APOSOR52669-1G APOSOR52669-250MG
2-Carboxypyridine-4-boronic acid, pinacol ester 95%
2-Carboxypyridine-4-boronic acid, pinacol ester
Formel: C₁₂H₁₆BNO₄
Molecular Weight: 249,071 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22192540
CAS-Nummer: 2096332-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR