3-Bromo-2-nitrobenzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

OR52706-10G OR52706-1G OR52706-5G
APOSOR52706-10GEA 656 EUR
APOSOR52706-10G APOSOR52706-1G APOSOR52706-5G
3-Bromo-2-nitrobenzaldehyde 97%
3-Bromo-2-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
Molecular Weight: 230,016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00462872
CAS-Nummer: 882722-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR