1-(6-Bromo-3-pyridyl)piperazine 95%

Lieferant: Apollo Scientific

OR52761-1G OR52761-500MG
APOSOR52761-1GEA 906 EUR
APOSOR52761-1G APOSOR52761-500MG
1-(6-Bromo-3-pyridyl)piperazine 95%
1-(6-Bromo-3-pyridyl)piperazine
Formel: C₉H₁₂BrN₃
Molecular Weight: 242.116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09910228
CAS-Nummer: 412347-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR