4-(6-Bromo-3-pyridyl)morpholine 98%

Lieferant: Apollo Scientific

Synonyme: 4-(6-Bromopyridin-3-yl)morpholine

OR52764-1G OR52764-250MG
APOSOR52764-1GEA 156 EUR
APOSOR52764-1G APOSOR52764-250MG
4-(6-Bromo-3-pyridyl)morpholine 98%
4-(6-Bromo-3-pyridyl)morpholine
Formel: C₉H₁₁BrN₂O
Molecular Weight: 243.1 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869572
CAS-Nummer: 952582-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR