2-(1-Piperidyl)pyridin-4-amine 95%

Lieferant: Apollo Scientific

OR52767-1G OR52767-250MG
APOSOR52767-1GEA 741 EUR
APOSOR52767-1G APOSOR52767-250MG
2-(1-Piperidyl)pyridin-4-amine 95%
2-(1-Piperidyl)pyridin-4-amine
Formel: C₁₀H₁₅N₃
Molecular Weight: 177,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03268586
CAS-Nummer: 35980-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR