3-(1-Piperidyl)pyridin-2-amine 95%

Lieferant: Apollo Scientific

OR52769-100MG OR52769-250MG
APOSOR52769-100MGEA 464 EUR
APOSOR52769-100MG APOSOR52769-250MG
3-(1-Piperidyl)pyridin-2-amine 95%
3-(1-Piperidyl)pyridin-2-amine
Formel: C₁₀H₁₅N₃
Molecular Weight: 177,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20727598
CAS-Nummer: 1286273-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR