6-Piperazin-1-ylpyridin-3-amine 98%
Lieferant: Apollo Scientific
OR52773-1G
OR52773-250MG
APOSOR52773-1GEA
683
EUR
APOSOR52773-1G
APOSOR52773-250MG
6-Piperazin-1-ylpyridin-3-amine 98%
6-Piperazin-1-ylpyridin-3-amine
Formel:
C₉H₁₄N₄ Molecular Weight: 178,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09882449 CAS-Nummer: 119285-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...