3-Morpholinopyridin-4-amine 95%

Lieferant: Apollo Scientific

OR52778-100MG OR52778-250MG
APOSOR52778-100MGEA 509 EUR
APOSOR52778-100MG APOSOR52778-250MG
3-Morpholinopyridin-4-amine 95%
3-Morpholinopyridin-4-amine
Formel: C₉H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD20715635
CAS-Nummer: 144864-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR