4-Acetyl-3-bromopyridine 97%
Lieferant: Apollo Scientific
Synonyme:
1-(3-Bromopyridin-4-yl)ethanone
OR52784-1G
OR52784-250MG
OR52784-5G
APOSOR52784-1GEA
232
EUR
APOSOR52784-1G
APOSOR52784-250MG
APOSOR52784-5G
4-Acetyl-3-bromopyridine 97%
4-Acetyl-3-bromopyridine
Formel:
C₇H₆BrNO Molecular Weight: 200,033 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128861 CAS-Nummer: 111043-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...