Methyl 2,6-dibromobenzoate 95%

Lieferant: Apollo Scientific

Synonyme: 2,6-Dibromobenzoic acid methyl ester

OR52900-1G OR52900-250MG OR52900-5G
APOSOR52900-1GEA 241 EUR
APOSOR52900-1G APOSOR52900-250MG APOSOR52900-5G
Methyl 2,6-dibromobenzoate 95%
Methyl 2,6-dibromobenzoate
Formel: C₈H₆Br₂O₂
Molecular Weight: 293,94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234397
CAS-Nummer: 873994-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR