Methyl 2-bromo-3,5-dinitrobenzoate 95%

Lieferant: Apollo Scientific

OR52919-1G OR52919-250MG OR52919-5G
APOSOR52919-1GEA 429 EUR
APOSOR52919-1G APOSOR52919-250MG APOSOR52919-5G
Methyl 2-bromo-3,5-dinitrobenzoate 95%
Methyl 2-bromo-3,5-dinitrobenzoate
Formel: C₈H₅BrN₂O₆
Molecular Weight: 305,039 g/mol
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD15466487
CAS-Nummer: 36749-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR