S-Acetyl-L-glutathione

Lieferant: Apollo Scientific

Synonyme: N5-((R)-3-(Acetylthio)-1-((carboxymethyl)amino)-1-oxopropan-2-yl)-L-glutamine

OR52950-25G
APOSOR52950-25GEA 120 EUR
APOSOR52950-25G
S-Acetyl-L-glutathione
S-Acetyl-L-glutathione
Formel: C₁₂H₁₉N₃O₇S
Molecular Weight: 349,36 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09952588
CAS-Nummer: 3054-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR