2-(Methoxymethylidene)propanedinitrile

Lieferant: Apollo Scientific

Synonyme: (Methoxymethylene)malononitrile

OR52959-1G OR52959-250MG OR52959-5G
APOSOR52959-1GEA 587 EUR
APOSOR52959-1G APOSOR52959-250MG APOSOR52959-5G
2-(Methoxymethylidene)propanedinitrile
2-(Methoxymethylidene)propanedinitrile
Formel: C₅H₄N₂O
Molecular Weight: 108,098 g/mol
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07437817
CAS-Nummer: 672-81-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR