6-Oxa-5-thia-4-azaspiro[2.4]heptane 5,5-dioxide, N-BOC protected
Lieferant: Apollo Scientific
Achtung
Synonyme: tert-Butyl 6-oxa-5-thia-4-azaspiro[2.4]heptane-4-carboxylate 5,5-dioxide
OR52985-1G OR52985-250MG
APOSOR52985-1GEA 433 EUR
APOSOR52985-1G APOSOR52985-250MG
6-Oxa-5-thia-4-azaspiro[2.4]heptane 5,5-dioxide, N-BOC protected
6-Oxa-5-thia-4-azaspiro[2.4]heptane 5,5-dioxide, N-BOC protected
Formel: C₉H₁₅NO₅S
Molecular Weight: 249.284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31925000
CAS-Nummer: 2023006-14-4