4-Bromo-3-(dimethylsulphamoyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR52989-1G OR52989-250MG
APOSOR52989-1GEA 248 EUR
APOSOR52989-1G APOSOR52989-250MG
4-Bromo-3-(dimethylsulphamoyl)benzoic acid
4-Bromo-3-(dimethylsulphamoyl)benzoic acid
Formel: C₉H₁₀BrNO₄S
Molecular Weight: 308,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06353544
CAS-Nummer: 473477-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR