4-Ethynyl-2-methylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethynyl-2-picoline, (2-Methylpyridin-4-yl)acetylene

OR53009-1G OR53009-250MG
APOSOR53009-1GEA 473 EUR
APOSOR53009-1G APOSOR53009-250MG
4-Ethynyl-2-methylpyridine
4-Ethynyl-2-methylpyridine
Formel: C₈H₇N
Molecular Weight: 117,148 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13175243
CAS-Nummer: 30413-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR