2-Ethynyl-5-methylpyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Ethynyl-3-picoline, (5-Methylpyridin-2-yl)acetylene
OR53010-1G
OR53010-250MG
APOSOR53010-1GEA
197
EUR
APOSOR53010-1G
APOSOR53010-250MG
2-Ethynyl-5-methylpyridine
2-Ethynyl-5-methylpyridine
Formel:
C₈H₇N Molecular Weight: 117.148 g/mol Siedepunkt: 97…99 °C (24 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD13175240 CAS-Nummer: 30413-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...