2-Chloro-3-ethynylpyrazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-3-ethynyl-1,4-diazine, (3-Chloropyrazin-2-yl)acetylene
OR53015-100MG
OR53015-500MG
APOSOR53015-100MGEA
65.4
EUR
APOSOR53015-100MG
APOSOR53015-500MG
2-Chloro-3-ethynylpyrazine
2-Chloro-3-ethynylpyrazine
Formel:
C₆H₃ClN₂ Molecular Weight: 138,554 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19689588 CAS-Nummer: 1374115-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...