2-Ethynyl-3-methylpyrazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Methylpyrazin-2-yl)acetylene, 2-Ethynyl-3-methyl-1,4-diazine
OR53020-100MG
OR53020-1G
OR53020-500MG
APOSOR53020-100MGEA
65.4
EUR
APOSOR53020-100MG
APOSOR53020-1G
APOSOR53020-500MG
2-Ethynyl-3-methylpyrazine 95+%
2-Ethynyl-3-methylpyrazine
Formel:
C₇H₆N₂ Molecular Weight: 118.136 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22199323 CAS-Nummer: 41039-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...