1H-imidazole-2-boronic acid hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Imidazol-2-yl)boronic acid hydrochloride

OR53072-250MG OR53072-1G OR53072-500MG
APOSOR53072-250MGEA 306 EUR
APOSOR53072-250MG APOSOR53072-1G APOSOR53072-500MG
1H-imidazole-2-boronic acid hydrochloride 95+%
1H-imidazole-2-boronic acid hydrochloride
Formel: C₃H₅BN₂O₂·ClH
Molecular Weight: 148.356 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16251134
CAS-Nummer: 1919022-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR