2'-Chloro-4'-bromoacetophenone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-2-chlorophenyl)ethanone, 1-(4-Bromo-2-chlorophenyl)ethan-1-one

OR53110-25G
APOSOR53110-25GEA 190 EUR
APOSOR53110-25G
2'-Chloro-4'-bromoacetophenone 97%
2'-Chloro-4'-bromoacetophenone
Formel: C₈H₆BrClO
Molecular Weight: 233,49 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781036
CAS-Nummer: 252561-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR