Piperidine-2-carbonitrile hemioxalate

Lieferant: Apollo Scientific
Achtung

OR53119-1G OR53119-5G
APOSOR53119-1GEA 321 EUR
APOSOR53119-1G APOSOR53119-5G
Piperidine-2-carbonitrile hemioxalate
Piperidine-2-carbonitrile hemioxalate
Formel: 2C₆H₁₀N₂·C₂H₂O₄
Molecular Weight: 310.349 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26959050
CAS-Nummer: 1523571-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR