Ethyl 2-hydrazinyl-1,3-thiazole-4-carboxylate hydrobromide

Lieferant: Apollo Scientific
Achtung

OR53130-1G OR53130-250MG
APOSOR53130-1GEA 406 EUR
APOSOR53130-1G APOSOR53130-250MG
Ethyl 2-hydrazinyl-1,3-thiazole-4-carboxylate hydrobromide
Ethyl 2-hydrazinyl-1,3-thiazole-4-carboxylate hydrobromide
Formel: C₆H₉N₃O₂S·BrH
Molecular Weight: 268.132 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30297372
CAS-Nummer: 67618-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR