4-Chloro-7-methoxyquinoline-6-carboxamide 97%

Lieferant: Apollo Scientific
Achtung

OR53151-5G OR53151-25G
APOSOR53151-5GEA 74.1 EUR
APOSOR53151-5G APOSOR53151-25G
4-Chloro-7-methoxyquinoline-6-carboxamide 97%
4-Chloro-7-methoxyquinoline-6-carboxamide
Formel: C₁₁H₉ClN₂O₂
Molecular Weight: 236,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192256
CAS-Nummer: 417721-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR