4-Chloropyrimidine hydrochloride 96%

Lieferant: Apollo Scientific
Achtung

OR53156-1G OR53156-5G
APOSOR53156-1GEA 119 EUR
APOSOR53156-1G APOSOR53156-5G
4-Chloropyrimidine hydrochloride 96%
4-Chloropyrimidine hydrochloride
Formel: C₄H₃ClN₂·ClH
Molecular Weight: 150,994 g/mol
Schmelzpunkt: 193…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11870807
CAS-Nummer: 179051-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR