Piperazin-2-one, N1-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-2-oxopiperazine, tert-Butyl 2-oxopiperazine-1-carboxylate
OR53157-1G
OR53157-5G
APOSOR53157-1GEA
285
EUR
APOSOR53157-1G
APOSOR53157-5G
Piperazin-2-one, N1-BOC protected
Piperazin-2-one, N1-BOC protected
Formel:
C₉H₁₆N₂O₃ Molecular Weight: 200,235 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06797597 CAS-Nummer: 889958-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...