1,4-Dihydro-6-iodo-4-oxoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 3-Carboxy-1,4-dihydro-6-iodo-4-oxoquinoline, 6-Iodo-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

OR54323-1G OR54323-500MG OR54323-5G
APOSOR54323-1GEA 219 EUR
APOSOR54323-1G APOSOR54323-500MG APOSOR54323-5G
1,4-Dihydro-6-iodo-4-oxoquinoline-3-carboxylic acid
1,4-Dihydro-6-iodo-4-oxoquinoline-3-carboxylic acid
Formel: C₁₀H₆INO₃
Molecular Weight: 315,064 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01912285
CAS-Nummer: 302949-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR