2,6-Dimethylisonicotinic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dimethylpyridine-4-carboxylic acid
OR54337-1G
OR54337-250MG
APOSOR54337-1GEA
132
EUR
APOSOR54337-1G
APOSOR54337-250MG
2,6-Dimethylisonicotinic acid 95%
2,6-Dimethylisonicotinic acid
Formel:
C₈H₉NO₂ Molecular Weight: 151,163 g/mol Schmelzpunkt: 276…291 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03731176 CAS-Nummer: 54221-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...