2,5-Dichloro-4-hydroxypyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dichloropyridin-4-ol
OR5435-1G
OR5435-250MG
OR5435-500MG
APOSOR5435-1GEA
241
EUR
APOSOR5435-1G
APOSOR5435-250MG
APOSOR5435-500MG
2,5-Dichloro-4-hydroxypyridine
2,5-Dichloro-4-hydroxypyridine
Formel:
C₅H₃Cl₂NO Molecular Weight: 163.989 g/mol Schmelzpunkt: 184 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01861982 CAS-Nummer: 847664-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...