5-Chloro-2-(chloromethyl)pyridine hydrochloride 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: α,5-Dichloro-2-picoline hydrochloride

OR54365-250MG OR54365-1G
APOSOR54365-250MGEA 154 EUR
APOSOR54365-250MG APOSOR54365-1G
5-Chloro-2-(chloromethyl)pyridine hydrochloride 96%
5-Chloro-2-(chloromethyl)pyridine hydrochloride
Formel: C₆H₅Cl₂N·ClH
Molecular Weight: 198.478 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20921510
CAS-Nummer: 124425-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR