4-Bromo-2-methylbenzyl bromide

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-(bromomethyl)toluene, 4-Bromo-1-(bromomethyl)-2-methylbenzene

OR54376-1G OR54376-250MG OR54376-5G
APOSOR54376-1GEA 202 EUR
APOSOR54376-1G APOSOR54376-250MG APOSOR54376-5G
4-Bromo-2-methylbenzyl bromide
4-Bromo-2-methylbenzyl bromide
Formel: C₈H₈Br₂
Molecular Weight: 263,957 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025011
CAS-Nummer: 156001-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR