Fmoc-(S)-4-aminopentanoic acid 99%

Lieferant: Apollo Scientific

Synonyme: (4S)-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid

OR54424-1G OR54424-250MG
APOSOR54424-1GEA 720 EUR
APOSOR54424-1G APOSOR54424-250MG
Fmoc-(S)-4-aminopentanoic acid 99%
Fmoc-(S)-4-aminopentanoic acid
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,385 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05663762
CAS-Nummer: 1014018-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR