N-(2-Aminoethyl)biotinamide

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2-Aminoethyl)-5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

OR54472-1G OR54472-250MG
APOSOR54472-1GEA 333 EUR
APOSOR54472-1G APOSOR54472-250MG
N-(2-Aminoethyl)biotinamide
N-(2-Aminoethyl)biotinamide
Formel: C₁₂H₂₂N₄O₂S
Molecular Weight: 286,394 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839905
CAS-Nummer: 1231488-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR