3-Bromo-2-nitrobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR54507-1G OR54507-5G OR54507-250MG
APOSOR54507-1GEA 60.9 EUR
APOSOR54507-1G APOSOR54507-5G APOSOR54507-250MG
3-Bromo-2-nitrobenzoic acid
3-Bromo-2-nitrobenzoic acid
Formel: C₇H₄BrNO₄
Molecular Weight: 246,015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10700064
CAS-Nummer: 116529-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR