(R)-3-Amino-4-methylpentanoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-D-β-HoVal-OH
OR54508-1G
OR54508-5G
APOSOR54508-1GEA
134
EUR
APOSOR54508-1G
APOSOR54508-5G
(R)-3-Amino-4-methylpentanoic acid 95%
(R)-3-Amino-4-methylpentanoic acid
Formel:
C₆H₁₃NO₂ Molecular Weight: 131,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01076231 CAS-Nummer: 75992-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...