8-Bromoisoquinolin-5-amine 95%

Lieferant: Apollo Scientific
Achtung

OR54531-1G OR54531-250MG
APOSOR54531-1GEA 1060 EUR
APOSOR54531-1G APOSOR54531-250MG
8-Bromoisoquinolin-5-amine 95%
8-Bromoisoquinolin-5-amine
Formel: C₉H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16621161
CAS-Nummer: 90721-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR