2,2-Dimethyl-3,4-dihydronaphthalen-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2-Dimethyl-3,4-dihydronaphthalen-1(2H)-one

OR54534-1G OR54534-5G
APOSOR54534-1GEA 63.8 EUR
APOSOR54534-1G APOSOR54534-5G
2,2-Dimethyl-3,4-dihydronaphthalen-1-one
2,2-Dimethyl-3,4-dihydronaphthalen-1-one
Formel: C₁₂H₁₄O
Molecular Weight: 174,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22565862
CAS-Nummer: 2977-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR