5-Bromo-7-methoxyquinoxaline

Lieferant: Apollo Scientific
Achtung

OR54563-1G OR54563-250MG
APOSOR54563-1GEA 638 EUR
APOSOR54563-1G APOSOR54563-250MG
5-Bromo-7-methoxyquinoxaline
5-Bromo-7-methoxyquinoxaline
Formel: C₉H₇BrN₂O
Molecular Weight: 239,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30481051
CAS-Nummer: 2149589-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR